ENA161208952 Enamine

1-(1H-benzimidazol-2-yl)ethanamine hydrochloride

  • MDL number MFCD08445279
  • Cas Number : 853789-10-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210...212
Purity 95.00
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02554

Safety Information

msds https://www.enaminestore.com/catalog/EN300-02554
Availability details for
Quantity
  • Availability
  • Add to Cart