ENA161208953 Enamine

2-[1-(piperazin-1-yl)ethyl]-1,3-benzothiazole dihydrochloride

  • MDL number MFCD07678493
  • Cas Number : 1052551-34-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115-116
Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13596

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13596
Availability details for
Quantity
  • Availability
  • Add to Cart