ENA161208958 Enamine

3-benzyl-1,3-thiazol-2(3H)-imine hydrochloride

  • MDL number MFCD00187833
  • Cas Number : 39913-88-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16...18
Purity 95.00
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13671

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-13671
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart