ENA161208964 Enamine

N-methyl-N-[(1-methyl-1H-imidazol-2-yl)methyl]amine dihydrochloride

  • MDL number MFCD07324813
  • Cas Number : 1049717-38-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206...208
Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13760

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13760
Availability details for
Quantity
  • Availability
  • Add to Cart