ENA161208966 Enamine

methyl 2-(chloromethyl)-4-phenylquinoline-3-carboxylate hydrochloride

  • MDL number MFCD07324820
  • Cas Number : 1052542-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 198...200
Purity 95.00
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13773

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13773
Availability details for
Quantity
  • Availability
  • Add to Cart