ENA161208973 Enamine

N-(2-methoxy-5-methylbenzyl)-N-methylamine hydrochloride

  • MDL number MFCD07323323
  • Cas Number : 1052550-44-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122...124
Purity 95.00
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13918

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13918
Availability details for
Quantity
  • Availability
  • Add to Cart