ENA161208982 Enamine

N-(4-sulfamoylphenyl)piperidine-4-carboxamide hydrochloride

  • MDL number MFCD08246122
  • Cas Number : 1052545-65-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 183-184
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14105

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14105
Availability details for
Quantity
  • Availability
  • Add to Cart