ENA161208986 Enamine

1-(piperidine-4-carbonyl)azepane hydrochloride

  • MDL number MFCD08445290
  • Cas Number : 1049723-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-14287

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14287
Availability details for
Quantity
  • Availability
  • Add to Cart