ENA161209000 Enamine

1-(4-chlorobenzyl)-1H-pyrazol-5-amine hydrochloride

  • MDL number MFCD07343853
  • Cas Number : 1052552-81-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 196...198
Purity 95.00
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14446

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14446
Availability details for
Quantity
  • Availability
  • Add to Cart