ENA161209039 Enamine

4-[(3,4-dimethylphenyl)sulfanyl]aniline hydrochloride

  • MDL number MFCD07284195
  • Cas Number : 1049762-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83-84
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06191

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06191
Availability details for
Quantity
  • Availability
  • Add to Cart