ENA161209040 Enamine

2-{[(thiophen-3-yl)methyl]sulfanyl}ethan-1-amine hydrochloride

  • MDL number MFCD07352248
  • Cas Number : 1173049-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179-180
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-14942

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14942
Availability details for
Quantity
  • Availability
  • Add to Cart