ENA161209042 Enamine

[8-chloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridin-3-yl]methanamine hydrochloride

  • MDL number MFCD07352246
  • Cas Number : 1052547-28-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-14959

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14959
Availability details for
Quantity
  • Availability
  • Add to Cart