ENA161209044 Enamine

methyl 3-(4-aminobenzenesulfonamido)propanoate hydrochloride

  • MDL number MFCD07355817
  • Cas Number : 1052546-14-1
  • Molecular Weight : 294.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148-149
Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-14965

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14965
Availability details for
Quantity
  • Availability
  • Add to Cart