ENA161209045 Enamine

1-[4-(piperidin-1-ylcarbonyl)phenyl]methanamine hydrochloride

  • MDL number MFCD07355818
  • Cas Number : 916762-48-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173...175
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14966

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14966
Availability details for
Quantity
  • Availability
  • Add to Cart