ENA161209053 Enamine

N-[2-hydroxy-5-(2-methyl-1,3-thiazol-4-yl)phenyl]acetamide hydrochloride

  • MDL number MFCD07681940
  • Cas Number : 1052550-99-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190...192
Purity 95.00
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15040

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15040
Availability details for
Quantity
  • Availability
  • Add to Cart