ENA161209066 Enamine

4-(piperazine-1-sulfonyl)benzene-1-sulfonamide hydrochloride

  • MDL number MFCD07685850
  • Cas Number : 1052543-23-3
  • Molecular Weight : 341.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 250-251
Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-15179

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15179
Availability details for
Quantity
  • Availability
  • Add to Cart