ENA161209067 Enamine

6-fluoro-3,4-dihydro-2H-1-benzothiopyran-4-amine hydrochloride

  • MDL number MFCD07685849
  • Cas Number : 1052544-66-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15181

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15181
Availability details for
Quantity
  • Availability
  • Add to Cart