ENA161209077 Enamine

3-(methylthio)-1-[1,2,4]triazolo[4,3-a]pyridin-3-ylpropan-1-amine dihydrochloride

  • MDL number MFCD07687285
  • Cas Number : 1049693-70-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165...167
Purity 95.00
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15271

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15271
Availability details for
Quantity
  • Availability
  • Add to Cart