ENA161209099 Enamine

ethyl 5-amino-1-(4-fluorophenyl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD00173917
  • Cas Number : 138907-68-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-16283

Safety Information

msds https://www.enaminestore.com/catalog/EN300-16283
Availability details for
Quantity
  • Availability
  • Add to Cart