ENA161209117 Enamine

4-(piperazine-1-sulfonyl)benzoic acid hydrochloride

  • MDL number MFCD07687254
  • Cas Number : 1043554-68-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 250-251
Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-21806

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-21806
Availability details for
Quantity
  • Availability
  • Add to Cart