ENA161209119 Enamine

N-ethyl-3,4-dihydro-2H-1,4-benzoxazine-2-carboxamide hydrochloride

  • MDL number MFCD08264945
  • Cas Number : 1049699-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 82-83
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-21812

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-21812
Availability details for
Quantity
  • Availability
  • Add to Cart