ENA161209122 Enamine

[4-(2-methyl-1,3-thiazol-4-yl)phenyl]methanamine dihydrochloride

  • MDL number MFCD07687100
  • Cas Number : 1052541-48-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195-196
Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-21826

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-21826
Availability details for
Quantity
  • Availability
  • Add to Cart