ENA161209131 Enamine

2-chloro-1-{4-[(thiophen-2-yl)methyl]piperazin-1-yl}ethan-1-one hydrochloride

  • MDL number MFCD07838434
  • Cas Number : 1049766-99-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-22600

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-22600
Availability details for
Quantity
  • Availability
  • Add to Cart