ENA161209138 Enamine

1-{4-[(5-bromothiophen-2-yl)methyl]piperazin-1-yl}-2-chloroethan-1-one hydrochloride

  • MDL number MFCD08692020
  • Cas Number : 1052544-19-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-22934

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-22934
Availability details for
Quantity
  • Availability
  • Add to Cart