ENA161209146 Enamine

2-chloro-1-{4-[(2-chloro-6-fluorophenyl)methyl]piperazin-1-yl}ethan-1-one hydrochloride

  • MDL number MFCD08448709
  • Cas Number : 1049738-89-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23017

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23017
Availability details for
Quantity
  • Availability
  • Add to Cart