ENA161209148 Enamine

methyl 3-amino-3-(3,4-dimethoxyphenyl)propanoate hydrochloride

  • MDL number MFCD06740360
  • Cas Number : 34841-03-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178...180
Purity 95.00
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23095

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23095
Availability details for
Quantity
  • Availability
  • Add to Cart