ENA161209149 Enamine

2-chloro-N-[2-methyl-2-(morpholin-4-yl)propyl]acetamide hydrochloride

  • MDL number MFCD09054652
  • Cas Number : 1171318-29-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-23109

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23109
Availability details for
Quantity
  • Availability
  • Add to Cart