ENA161209153 Enamine

N-(1-benzylpyrrolidin-3-yl)-2-chloroacetamide hydrochloride

  • MDL number MFCD09373424
  • Cas Number : 1170955-45-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23163

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23163
Availability details for
Quantity
  • Availability
  • Add to Cart