ENA161209160 Enamine

isopropyl 3-amino-3-phenylpropanoate hydrochloride

  • MDL number MFCD07838441
  • Cas Number : 100369-81-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23275

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23275
Availability details for
Quantity
  • Availability
  • Add to Cart