ENA161209162 Enamine

N-ethylpiperazine-1-carboxamide hydrochloride

  • MDL number MFCD08246125
  • Cas Number : 474711-90-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-23396

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23396
Availability details for
Quantity
  • Availability
  • Add to Cart