ENA161209163 Enamine

methyl 3-amino-3-(4-methylphenyl)propanoate hydrochloride

  • MDL number MFCD09834089
  • Cas Number : 101207-50-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23416

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23416
Availability details for
Quantity
  • Availability
  • Add to Cart