ENA161209171 Enamine

3-methyl-4-(4-methylpiperazin-1-yl)aniline dihydrochloride

  • MDL number MFCD07850242
  • Cas Number : 1177349-04-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104...106
Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23514

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23514
Availability details for
Quantity
  • Availability
  • Add to Cart