ENA161209176 Enamine

N-(2-aminoethyl)-N-methyl-N-phenylamine dihydrochloride

  • MDL number MFCD07841700
  • Cas Number : 36271-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 212...214
Purity 95.00
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23578

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23578
Availability details for
Quantity
  • Availability
  • Add to Cart