ENA161209182 Enamine

2-amino-2-[1-(3-chlorophenyl)-3,5-dimethyl-1H-pyrazol-4-yl]acetic acid hydrochloride

  • MDL number MFCD07838423
  • Cas Number : 1078161-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-23659

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23659
Availability details for
Quantity
  • Availability
  • Add to Cart