ENA161209192 Enamine

1-[(dimethyl-1,2-oxazol-4-yl)sulfonyl]-1,4-diazepane hydrochloride

  • MDL number MFCD07838468
  • Cas Number : 1049775-82-7
  • Molecular Weight : 295.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-23810

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23810
Availability details for
Quantity
  • Availability
  • Add to Cart