ENA161209208 Enamine

1-[2-(aminomethyl)phenyl]-N-isopropylmethanesulfonamide hydrochloride

  • MDL number MFCD08239601
  • Cas Number : 1052089-42-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24169

Safety Information

msds https://www.enaminestore.com/catalog/EN300-24169
Availability details for
Quantity
  • Availability
  • Add to Cart