ENA161209209 Enamine

[4-(aminomethyl)phenyl]methanesulfonamide hydrochloride

  • MDL number MFCD08239598
  • Cas Number : 854637-10-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-24170

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24170
Availability details for
Quantity
  • Availability
  • Add to Cart