ENA161209210 Enamine

N-phenylpiperidine-3-carboxamide hydrochloride

  • MDL number MFCD08246137
  • Cas Number : 521969-45-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 226...228
Purity 95.00
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24187

Safety Information

msds https://www.enaminestore.com/catalog/EN300-24187
Availability details for
Quantity
  • Availability
  • Add to Cart