ENA161209218 Enamine

4-amino-3-(methylsulfanyl)benzoic acid hydrochloride

  • MDL number MFCD08262845
  • Cas Number : 1049711-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-24332

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24332
Availability details for
Quantity
  • Availability
  • Add to Cart