ENA161209232 Enamine

4-[2-(4-methoxyphenyl)ethyl]piperidine hydrochloride

  • MDL number MFCD08456918
  • Cas Number : 148135-92-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24696

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24696
Availability details for
Quantity
  • Availability
  • Add to Cart