ENA161209240 Enamine

3-(3-cyano-5,7-dimethylpyrazolo[1,5-a]pyrimidin-6-yl)propanoic acid

  • MDL number MFCD08166736
  • Cas Number : 900137-38-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25414

Safety Information

msds https://www.enaminestore.com/catalog/EN300-25414
Availability details for
Quantity
  • Availability
  • Add to Cart