ENA161209254 Enamine

2-(1H-imidazol-2-yl)-1-phenylethan-1-one hydrochloride

  • MDL number MFCD07701064
  • Cas Number : 52855-75-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25760

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25760
Availability details for
Quantity
  • Availability
  • Add to Cart