ENA161209257 Enamine

1-{[2-(thiophen-3-yl)-1,3-thiazol-4-yl]methyl}piperazine hydrochloride

  • MDL number MFCD08692018
  • Cas Number : 1052539-68-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 192-193
Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26010

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26010
Availability details for
Quantity
  • Availability
  • Add to Cart