ENA161209258 Enamine

1-[(2-methyl-1,3-thiazol-4-yl)methyl]piperazine hydrochloride

  • MDL number MFCD07771967
  • Cas Number : 1052539-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26016

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-26016
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart