ENA161209267 Enamine

1-{[3-(4-methoxyphenyl)-1,2,4-oxadiazol-5-yl]methyl}piperazine hydrochloride

  • MDL number MFCD08692026
  • Cas Number : 1052553-32-8
  • Molecular Weight : 310.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26211

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26211
Availability details for
Quantity
  • Availability
  • Add to Cart