ENA161209268 Enamine

ethyl 2-(4-chlorophenyl)-2-(ethylamino)acetate hydrochloride

  • MDL number MFCD08692035
  • Cas Number : 1046535-04-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26275

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26275
Availability details for
Quantity
  • Availability
  • Add to Cart