ENA161209269 Enamine

1-{[2-(4-methoxyphenyl)-1,3-thiazol-4-yl]methyl}piperazine hydrochloride

  • MDL number MFCD08692021
  • Cas Number : 1052544-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Salt Data HCl
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26289

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26289
Availability details for
Quantity
  • Availability
  • Add to Cart