ENA161209270 Enamine

6-methyl-3,6,7,8-tetrahydro-2H-spiro[[1,4]dioxino[2,3-g]isoquinoline-9,1'-cyclopentane] hydrochloride

  • MDL number MFCD09296975
  • Cas Number : 153643-98-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26291

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26291
Availability details for
Quantity
  • Availability
  • Add to Cart