ENA161209290 Enamine

3-(sulfanylmethyl)benzoic acid

  • MDL number MFCD10686660
  • Cas Number : 28162-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104-106
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26698

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26698
Availability details for
Quantity
  • Availability
  • Add to Cart