ENA161209306 Enamine

4-methyl-5-(piperazine-1-sulfonyl)-2,3-dihydro-1,3-thiazol-2-one hydrochloride

  • MDL number MFCD09054660
  • Cas Number : 1170915-85-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27118

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27118
Availability details for
Quantity
  • Availability
  • Add to Cart